Triethylsilane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10320-09
A10320-18
A10320-30
CAAAA10320-09EA
58.62
CAD
CAAAA10320-09
CAAAA10320-18
CAAAA10320-30
Triethylsilane 98+%
Triethylsilane
MDL: MFCD00009018
Beilstein Registry No.: 1098278
Fieser: 1,1218 15,338 16,356 17,367 18,373 19,360 20,188 21,218
Purity: 98+%
CAS No.: 617-86-7
Molecular Formula: C6H16Si
Formula Weight: 116.28
Flash Point: -6°(21°F)
Refractive Index: 1.4120
Melting Point: -157°
Boiling Point: 107-108°
Beilstein Reference: 1098278
UN No.: 1993
MDL No.: MFCD00009018
Formula:
C₆H₁₆Si MW: 116.28 g/mol Boiling Pt: 107 Melting Pt: -156.9 Density: 0.728 Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009018 CAS Number: 617-86-7 EINECS: 210-535-3 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...