4-Bromo-1,2-dichlorobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

A10314-06 A10314-14 A10314-22
CAAAA10314-06EA 47.72 CAD
CAAAA10314-06 CAAAA10314-14 CAAAA10314-22
4-Bromo-1,2-dichlorobenzene 98+%
4-Bromo-1,2-dichlorobenzene

MDL: MFCD00040849 Beilstein Registry No.: 1635735


Purity: 98+% CAS No.: 18282-59-2 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Flash Point: 87°(188°F) Melting Point: 22-25° Boiling Point: 140-142°/30mm Beilstein Reference: 1635735 MDL No.: MFCD00040849

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 140…142 °C/30 mm
Melting Pt: 22…25 °C
Density: 1.781
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR