cis-3-Hexen-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10313-09
A10313-18
A10313-30
CAAAA10313-09EA
72.52
CAD
CAAAA10313-09
CAAAA10313-18
CAAAA10313-30
cis-3-Hexen-1-ol 98%
cis-3-Hexen-1-ol
MDL: MFCD00063217
Beilstein Registry No.: 1719712
Purity: 98%
CAS No.: 928-96-1
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 44°(111°F)
Refractive Index: 1.4400
Odor: Pleasant
Boiling Point: 156-157°
Beilstein Reference: 1719712
Merck Reference: 14,4700
UN No.: 1987
MDL No.: MFCD00063217
Formula:
C₆H₁₂O Boiling Pt: 156…157 °C Melting Pt: –60 °C Density: 0.848 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063217 CAS Number: 928-96-1 EINECS: 213-192-8 UN: 1987 ADR: 3,III Merck Index: 14,4700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...