N-Hydroxysuccinimide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
A10312-14
A10312-22
A10312-36
CAAAA10312-14EA
75.37
CAD
CAAAA10312-14
CAAAA10312-22
CAAAA10312-36
N-Hydroxysuccinimide 98+%
N-Hydroxysuccinimide
MDL: MFCD00005516
Beilstein Registry No.: 113913
Fieser: 1,487 9,246
Purity: 98+%
CAS No.: 6066-82-6
Molecular Formula: C4H5NO3
Formula Weight: 115.09
Melting Point: 94-99°
Beilstein Reference: 113913
Formula:
C₄H₅NO₃ MW: 115.09 g/mol Boiling Pt: 363 °C (1013 hPa) Melting Pt: 94…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005516 CAS Number: 6066-82-6 EINECS: 228-001-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...