2-Chlorophenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10311-0B
A10311-0I
A10311-30
CAAAA10311-0BEA
131.14
CAD
CAAAA10311-0B
CAAAA10311-0I
CAAAA10311-30
2-Chlorophenol 99%
2-Chlorophenol
MDL: MFCD00002159
Beilstein Registry No.: 1905114
Purity: 99%
CAS No.: 95-57-8
Molecular Formula: C6H5ClO
Formula Weight: 128.56
Flash Point: 63°(145°F)
Refractive Index: 1.5590
Odor: Phenol-like
Melting Point: 7-8°
Boiling Point: 175-176°
Beilstein Reference: 1905114
Merck Reference: 14,2154
UN No.: 2021
MDL No.: MFCD00002159
Formula:
C₆H₅ClO Boiling Pt: 175…176 °C Melting Pt: 7…8 °C Density: 1.260 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002159 CAS Number: 95-57-8 EINECS: 202-433-2 UN: 2021 ADR: 6.1,III Merck Index: 14,2154 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...