ε-Hexalactone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
ε-Caprolactone, Oxepan-2-one, Hexan-6-olide
A10299-22
A10299-0C
A10299-0E
A10299-36
CAAAAA10299-22EA
42.98
CAD
CAAAAA10299-22
CAAAA10299-0C
CAAAA10299-0E
CAAAA10299-36
ε-Hexalactone 99%
ε-Hexalactone
MDL: MFCD00003267
Beilstein Registry No.: 106919
Purity: 99%
CAS No.: 502-44-3
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 109°(228°F)
Physical Form: Liquid
Density: 1.078
Refractive Index: 1.4630
Appearance: Colorless
Melting Point: -2°
Boiling Point: 235-236°
Beilstein Reference: 106919
MDL No.: MFCD00003267
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 235…236 °C Melting Pt: –2 °C Density: 1.078 Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003267 CAS Number: 502-44-3 EINECS: 207-938-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...