Ethyl isovalerate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10292-0F
A10292-AE
A10292-AP
CAAAA10292-0FEA
336.54
CAD
CAAAA10292-0F
CAAAA10292-AE
CAAAA10292-AP
Ethyl isovalerate 98%
Ethyl isovalerate
MDL: MFCD00009203
Beilstein Registry No.: 1744677
Purity: 98%
CAS No.: 108-64-5
Molecular Formula: (CH3)2CHCH2CO2CH2CH3
Formula Weight: 130.19
Flash Point: 26°(78°F)
Refractive Index: 1.3960
Odor: Fruit-like
Melting Point: -99°
Boiling Point: 131-133°
Beilstein Reference: 1744677
Merck Reference: 14,3816
UN No.: 3272
MDL No.: MFCD00009203
Formula:
C₇H₁₄O₂ Boiling Pt: 131…133 °C Melting Pt: –99 °C Density: 0.866 Flash Pt: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009203 CAS Number: 108-64-5 EINECS: 203-602-3 UN: 1993 ADR: 3,III Merck Index: 14,3816 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...