3,3-Dimethylbutanone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10289-22
A10289-36
CAAAA10289-22EA
165.9
CAD
CAAAA10289-22
CAAAA10289-36
3,3-Dimethylbutanone 97%
3,3-Dimethylbutanone
MDL: MFCD00008846
Purity: 97%
CAS No.: 75-97-8
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 5°(41°F)
Refractive Index: 1.3970
Melting Point: -53°
Boiling Point: 105-106°
Merck Reference: 14,7440
UN No.: 1224
MDL No.: MFCD00008846
Formula:
C₆H₁₂O Boiling Pt: 105…106 °C Melting Pt: –53 °C Density: 0.803 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008846 CAS Number: 75-97-8 EINECS: 200-920-4 UN: 1224 ADR: 3,III Merck Index: 14,7440 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...