o-Anisidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Anisidine, 2-Aminoanisole, 2-Methoxyaniline

A10288-0B A10288-0I A10288-30
CAAAA10288-0BEA 195.92 CAD
CAAAA10288-0B CAAAA10288-0I CAAAA10288-30
o-Anisidine 99%
o-Anisidine

MDL: MFCD00007688 Beilstein Registry No.: 386210


Purity: 99% CAS No.: 90-04-0 Molecular Formula: C7H9NO Formula Weight: 123.16 Flash Point: 98°(208°F) Physical Form: Liquid Density: 1.092 Refractive Index: 1.5740 Sensitivity: Light Sensitive Appearance: Yellow to brown Melting Point: 5-6° Boiling Point: 224-225° Beilstein Reference: 386210 Merck Reference: 14,667 UN No.: 2431 MDL No.: MFCD00007688

Formula: C₇H₉NO
Boiling Pt: 224…225 °C
Melting Pt: 5…6 °C
Density: 1.092
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007688
CAS Number: 90-04-0
EINECS: 201-963-1
UN: 2431
ADR: 6.1,III
Merck Index: 14,667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR