Isonicotinonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine
A10286-09
A10286-0E
A10286-22
A10286-36
CAAAAA10286-09EA
37.6
CAD
CAAAAA10286-09
CAAAA10286-0E
CAAAA10286-22
CAAAA10286-36
Isonicotinonitrile 98%
Isonicotinonitrile
MDL: MFCD00006417
Beilstein Registry No.: 107712
Purity: 98%
CAS No.: 100-48-1
Molecular Formula: C6H4N2
Formula Weight: 104.11
Flash Point: 88°(190°F)
Odor: Pungent
pH: 6
Melting Point: 76-80°
Boiling Point: 194-196°
Beilstein Reference: 107712
MDL No.: MFCD00006417
Formula:
C₆H₄N₂ Boiling Pt: 194…196 °C Melting Pt: 76…80 °C Flash Pt: 88 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006417 CAS Number: 100-48-1 EINECS: 202-856-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...