Dichloro(phenyl)phosphine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10284-0B
A10284-18
A10284-30
CAAAA10284-0BEA
334.96
CAD
CAAAA10284-0B
CAAAA10284-18
CAAAA10284-30
Dichloro(phenyl)phosphine 97%
Dichloro(phenyl)phosphine
MDL: MFCD00000528
Beilstein Registry No.: 508189
Purity: 97%
CAS No.: 644-97-3
Molecular Formula: C6H5Cl2P
Formula Weight: 178.99
Flash Point: 102°(215°F)
Refractive Index: 1.5980
Odor: Pungent
Melting Point: -51°
Boiling Point: 99-101°/11mm
Beilstein Reference: 508189
UN No.: 2798
MDL No.: MFCD00000528
Formula:
C₆H₅Cl₂P Boiling Pt: 99…101 °C/11 mm Melting Pt: –51 °C Density: 1.319 Flash Pt: 102 °C (215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000528 CAS Number: 644-97-3 EINECS: 211-425-8 UN: 2798 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...