3-Chlorobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Chlorobenzene-1-sulfonyl chloride, 3-Chlorobenzene-1-sulphonyl chloride

A10283-03 A10283-06 A10283-14
CAAAA10283-03EA 51.98 CAD
CAAAA10283-03 CAAAA10283-06 CAAAA10283-14
3-Chlorobenzenesulfonyl chloride 98%
3-Chlorobenzenesulphonyl chloride

MDL: MFCD00051697 Beilstein Registry No.: 2937139


Purity: 98% CAS No.: 2888-06-4 Molecular Formula: C6H4Cl2O2S Formula Weight: 211.07 Flash Point: 110°(230°F) Refractive Index: 1.5690 Melting Point: ca 21° Boiling Point: 102-104°/1mm Beilstein Reference: 2937139 UN No.: 3265 MDL No.: MFCD00051697

Formula: C₆H₄Cl₂O₂S
MW: 211.07
Boiling Pt: 102…104 °C (1 mmHg)
Melting Pt: ∼ 21 °C
Density: 1.499
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051697
CAS Number: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR