Iron(III) citrate hydrate Fe(III) 16.5-20% (Fe(II) ≤ 5%)

Supplier: Thermo Scientific Chemicals

Synonyms: Ferric citrate

A10276-0B A10276-18 A10276-30
CAAAA10276-0BEA 244.9 CAD
CAAAA10276-0B CAAAA10276-18 CAAAA10276-30
Iron(III) citrate hydrate Fe(III) 16.5-20% (Fe(II) ≤ 5%)
Iron(III) citrate hydrate

Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.


CAS No.: 334024-15-6 Molecular Formula: C6H5FeO7.xH2O Formula Weight: 244.94(anhy) MDL No.: MFCD00149617

Formula: C₆H₅FeO₇·xH₂O
MW: 244.948 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149615
CAS Number: 334024-15-6
EINECS: 222-536-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR