2-Bromoethanol 97%

Supplier: Thermo Scientific Chemicals
Danger

A10275-14 A10275-0B A10275-30
CAAAAA10275-14EA 60.36 CAD
CAAAAA10275-14 CAAAA10275-0B CAAAA10275-30
2-Bromoethanol 97%
2-Bromoethanol

MDL: MFCD00002827 Beilstein Registry No.: 878140 Fieser: 11,77


Purity: 97% CAS No.: 540-51-2 Molecular Formula: C2H5BrO Formula Weight: 124.97 Flash Point: 110°(230°F) Refractive Index: 1.4930 Melting Point: -80° Boiling Point: 149-150° Beilstein Reference: 878140 Merck Reference: 14,3792 UN No.: 2922 MDL No.: MFCD00002827

Formula: BrH₂CCH₂OH
Boiling Pt: 149…150 °C
Melting Pt: –80 °C
Density: 1.762
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002827
CAS Number: 540-51-2
EINECS: 208-748-1
UN: 2927
ADR: 6.1,II
Merck Index: 14,3792

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR