1-Naphthoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Naphthoic acid

A10273-14 A10273-22
CAAAA10273-14EA 49.77 CAD
CAAAA10273-14 CAAAA10273-22
1-Naphthoic acid 98%
1-Naphthoic acid

MDL: MFCD00004007 Beilstein Registry No.: 1908896


Purity: 98% CAS No.: 86-55-5 Molecular Formula: C11H8O2 Formula Weight: 172.18 Flash Point: 195°(383°F) Melting Point: 160-162° Boiling Point: 300° Beilstein Reference: 1908896 Merck Reference: 14,6381 MDL No.: MFCD00004007

Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: >300 °C
Melting Pt: 160…162 °C
Density: 1.398
Flash Pt: 195 °C(383 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004007
CAS Number: 86-55-5
EINECS: 201-681-9
Merck Index: 14,6381

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR