2,5-Difluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10272-09
A10272-18
CAAAA10272-09EA
76.31
CAD
CAAAA10272-09
CAAAA10272-18
2,5-Difluoroaniline 98%
2,5-Difluoroaniline
MDL: MFCD00007651
Beilstein Registry No.: 2802549
Purity: 98%
CAS No.: 367-30-6
Molecular Formula: C6H5F2N
Formula Weight: 129.11
Flash Point: 68°(154°F)
Refractive Index: 1.5130
Odor: Characteristic
Melting Point: 13-14°
Boiling Point: 171-174°
Beilstein Reference: 2802549
UN No.: 2941
MDL No.: MFCD00007651
Formula:
C₆H₅F₂N Boiling Pt: 171…174 °C Melting Pt: 13…14 °C Density: 1.285 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007651 CAS Number: 367-30-6 EINECS: 206-690-1 UN: 2941 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...