Trimethyl orthobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10270-09
A10270-18
A10270-30
CAAAA10270-09EA
69.52
CAD
CAAAA10270-09
CAAAA10270-18
CAAAA10270-30
Trimethyl orthobenzoate 98%
Trimethyl orthobenzoate
MDL: MFCD00008474
Beilstein Registry No.: 777623
Purity: 98%
CAS No.: 707-07-3
Molecular Formula: (C6H5)C(OCH3)3
Formula Weight: 182.22
Flash Point: 82°(179°F)
Refractive Index: 1.4890
Boiling Point: 87-88°/7mm
Beilstein Reference: 777623
MDL No.: MFCD00008474
Formula:
C₁₀H₁₄O₃ Boiling Pt: 87…88 °C/7 mm Density: 1.060 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008474 CAS Number: 707-07-3 EINECS: 211-897-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...