5-Chloro-2-pentanone 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10269-22
A10269-36
CAAAA10269-22EA
156.1
CAD
CAAAA10269-22
CAAAA10269-36
5-Chloro-2-pentanone 94%
5-Chloro-2-pentanone
MDL: MFCD00001008
Beilstein Registry No.: 1098490
Purity: 94%
CAS No.: 5891-21-4
Molecular Formula: C5H9ClO
Formula Weight: 120.58
Storage Temperature: 2°C to 8°C
Flash Point: 35°(95°F)
Refractive Index: 1.4380
Boiling Point: 63-64°/12mm
Beilstein Reference: 1098490
UN No.: 1224
MDL No.: MFCD00001008
Formula:
C₅H₉ClO Boiling Pt: 63…64 °C/12 mm Melting Pt: –48 °C Density: 1.053 Flash Pt: 35 °C (95 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001008 CAS Number: 5891-21-4 EINECS: 227-565-8 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...