Tributyl citrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Citric acid tributyl ester, Hydroxytricarballylic acid tributyl ester, Tributyl 2-hydroxypropane-1,2,3-tricarboxylate

A10266-0B A10266-30
CAAAA10266-0BEA 300.2 CAD
CAAAA10266-0B CAAAA10266-18 CAAAA10266-30
Tributyl citrate 99%
Tributyl citrate

MDL: MFCD00027217 Beilstein Registry No.: 1806072


Purity: 99% CAS No.: 77-94-1 Molecular Formula: C18H32O7 Formula Weight: 360.45 Flash Point: 157°(315°F) Refractive Index: 1.4440 Odor: Weak Melting Point: -20° Boiling Point: 325° Beilstein Reference: 1806072 Merck Reference: 14,1564 MDL No.: MFCD00027217

Formula: C₁₈H₃₂O₇
MW: 360.45 g/mol
Boiling Pt: 325 °C
Melting Pt: –20 °C
Density: 1.040
Flash Pt: 157 °C(315 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027217
CAS Number: 77-94-1
EINECS: 201-071-2
Merck Index: 14,1564

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR