5-Nitrosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-5-nitrobenzaldehyde

A10264-22 A10264-06 A10264-14
CAAAAA10264-22EA 641.48 CAD
CAAAAA10264-22 CAAAA10264-06 CAAAA10264-14
5-Nitrosalicylaldehyde 98%
5-Nitrosalicylaldehyde

MDL: MFCD00007337 Beilstein Registry No.: 512565


Purity: 98% CAS No.: 97-51-8 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 123-127° Beilstein Reference: 512565

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00007337
CAS Number: 97-51-8
EINECS: 202-587-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR