di-Ammonium oxalate monohydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10263-0E
A10263-22
A10263-36
CAAAA10263-0EEA
438.69
CAD
CAAAA10263-0E
CAAAA10263-22
CAAAA10263-36
di-Ammonium oxalate monohydrate 98%
di-Ammonium oxalate monohydrate
MDL: MFCD00149694
Beilstein Registry No.: 3759830
Purity: 98%
CAS No.: 6009-70-7
Molecular Formula: C2H8N2O4.H2O
Formula Weight: 142.11 (124.10anhy)
Physical Form: Crystalline solid
Density: 1.50
Appearance: White
Melting Point: 70° dec.
Beilstein Reference: 3759830
Merck Reference: 14,537
UN No.: 3288
MDL No.: MFCD00149694
Formula:
(NH₄)₂C₂O₄·H₂O Boiling Pt: 70 °C (1013 hPa) Melting Pt: 70 °C (dec.) Density: 1.50 Storage Temperature: Ambient |
MDL Number:
MFCD00149694 CAS Number: 6009-70-7 EINECS: 238-135-4 UN: 3288 ADR: 6.1,III Merck Index: 14,537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...