2-Oxoglutaric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid
A10256-14
A10256-22
A10256-36
CAAAA10256-14EA
55.93
CAD
CAAAA10256-14
CAAAA10256-22
CAAAA10256-36
2-Oxoglutaric acid 98%
2-Oxoglutaric acid
MDL: MFCD00004165
Beilstein Registry No.: 1705689
Fieser: 1,531
Purity: 98%
CAS No.: 328-50-7
Molecular Formula: C5H6O5
Formula Weight: 146.10
Storage Temperature: 2°C to 8°C
Melting Point: 113-117°
Beilstein Reference: 1705689
Merck Reference: 14,5303
MDL No.: MFCD00004165
Formula:
HOOCCH₂CH₂COCOOH MW: 146.1 g/mol Boiling Pt: 323 °C (1013 hPa) Melting Pt: 113…117 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004165 CAS Number: 328-50-7 EINECS: 206-330-3 Merck Index: 14,5303 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...