3-Methyl-1,2-cyclopentanedione (2-hydroxy-3-methylcyclopent-2-enone, methyl cyclopentenolone) 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Methyl-1,2-cyclopentanedione, Maple lactone, Methyl cyclopentenolone, 3-Methylcyclopentane-1,2-dione

A10255-09 A10255-18 A10255-22
CAAAAA10255-09EA 47.56 CAD
CAAAAA10255-09 CAAAAA10255-18 CAAAA10255-22
3-Methyl-1,2-cyclopentanedione (2-hydroxy-3-methylcyclopent-2-enone, methyl cyclopentenolone) 98+%
3-Methyl-1,2-cyclopentanedione

MDL: MFCD00001417 Beilstein Registry No.: 2076520

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001417
CAS Number: 765-70-8
EINECS: 212-154-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR