Ethyl-3-bromobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10251-09 A10251-18
CAAAA10251-09EA 93.38 CAD
CAAAA10251-09 CAAAA10251-18 CAAAA10251-30
Ethyl-3-bromobenzoate 98+%
Ethyl 3-bromobenzoate

MDL: MFCD00013529 Beilstein Registry No.: 1867121


Purity: 98+% CAS No.: 24398-88-7 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Flash Point: 110°(230°F) Refractive Index: 1.5430 Boiling Point: 261° Beilstein Reference: 1867121 MDL No.: MFCD00013529

Formula: C₉H₉BrO₂
Boiling Pt: 261 °C
Density: 1.430
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013529
CAS Number: 24398-88-7
EINECS: 246-223-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR