4-Pentylbenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10248-06
A10248-14
A10248-22
CAAAA10248-06EA
124.66
CAD
CAAAA10248-06
CAAAA10248-14
CAAAA10248-22
4-Pentylbenzoyl chloride 98%
4-Pentylbenzoyl chloride
MDL: MFCD00000700
Beilstein Registry No.: 2207887
Purity: 98%
CAS No.: 49763-65-7
Molecular Formula: C12H15ClO
Formula Weight: 210.71
Flash Point:
Formula:
C₁₂H₁₅ClO Boiling Pt: 144…145 °C/10 mm Density: 1.034 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000700 CAS Number: 49763-65-7 EINECS: 256-478-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...