D-Alanine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
A10231-06
A10231-14
A10231-22
CAAAA10231-06EA
61.94
CAD
CAAAA10231-06
CAAAA10231-14
CAAAA10231-22
D-Alanine 99%
D-Alanine
MDL: MFCD00008077
Beilstein Registry No.: 1720249
Optical Rotation: -14°C (c=5 in 5N HCl)
Purity: 99%
CAS No.: 338-69-2
Molecular Formula: C3H7NO2
Formula Weight: 89.09
Melting Point: ca 295° dec.
Beilstein Reference: 1720249
Merck Reference: 14,204
Optical Rotation: -14° (c=5 in 5N HCl)
MDL No.: MFCD00008077
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 189 Melting Pt: 79 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008077 CAS Number: 338-69-2 EINECS: 206-418-1 Merck Index: 14,204 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...