L(-)-Tryptophan 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
A10230-14
A10230-22
A10230-36
CAAAA10230-14EA
74.42
CAD
CAAAA10230-14
CAAAA10230-22
CAAAA10230-36
L(-)-Tryptophan 99%
L(-)-Tryptophan
MDL: MFCD00064340
Beilstein Registry No.: 86197
Optical Rotation: -31.5°C (c=1 in water)
Purity: 99%
CAS No.: 73-22-3
Molecular Formula: C11H12N2O2
Formula Weight: 204.23
Melting Point: 280° 285°
Beilstein Reference: 86197
Merck Reference: 14,9797
Optical Rotation: -31° (c=1 in water)
MDL No.: MFCD00064340
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 280 °C 285 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 Merck Index: 14,9797 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...