2-Amino-3-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline

A10224-14 A10224-22
CAAAAA10224-14EA 64.62 CAD
CAAAAA10224-14 CAAAA10224-22 CAAAA10224-36
2-Amino-3-methylpyridine 97%
2-Amino-3-methylpyridine

MDL: MFCD00006320 Beilstein Registry No.: 107892


Purity: 97% CAS No.: 1603-40-3 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 118°(244°F) Refractive Index: 1.5825 Odor: Unpleasant Melting Point: 29-32° Boiling Point: 221-222° Beilstein Reference: 107892 UN No.: 2811 MDL No.: MFCD00006320

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 221.1
Melting Pt: 29…32 °C
Density: 1.073
Flash Pt: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR