Imidazole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10221-0E
A10221-22
A10221-36
CAAAA10221-0EEA
625.68
CAD
CAAAA10221-0E
CAAAA10221-22
CAAAA10221-36
Imidazole 99%
Imidazole
Excellent for buffer use in pH range 6.2 - 7.8. Also an histamine antagonist.
Purity: 99%
CAS No.: 288-32-4
Molecular Formula: C3H4N2
Formula Weight: 68.08
Flash Point: 145°(293°F)
Physical Form: Cryst flakes
Density: 1.03
Appearance: White to cream
Melting Point: 88-92°
Boiling Point: 256°
Beilstein Reference: 103853
Merck Reference: 14,4912
UN No.: 2923
MDL No.: MFCD00005183
Excellent for buffer use in pH range 6.2 - 7.8. Also an histamine antagonist.
Formula:
C₃H₄N₂ Boiling Pt: 256 °C Melting Pt: 88…92 °C Density: 1.03 Flash Pt: 145 °C (293 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 EINECS: 206-019-2 UN: 3263 ADR: 8,III Merck Index: 14,4912 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...