2',6'-Difluoroacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2,6-Difluorophenyl)ethan-1-one, 1-(2,6-Difluorophenyl)ethanone
A10220-06
A10220-14
CAAAA10220-03EA
134.19
CAD
CAAAA10220-03
CAAAA10220-06
CAAAA10220-14
2',6'-Difluoroacetophenone 98%
2',6'-Difluoroacetophenone
MDL: MFCD00000328
Beilstein Registry No.: 2046068
Purity: 98%
CAS No.: 13670-99-0
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Flash Point: 76°(168°F)
Refractive Index: 1.4810
Boiling Point: 76-79°/15mm
Beilstein Reference: 2046068
MDL No.: MFCD00000328
Formula:
C₈H₆F₂O MW: 156.13 g/mol Boiling Pt: 76…79 °C/15 mm Density: 1,195 Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000328 CAS Number: 13670-99-0 EINECS: 237-151-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...