2,3-Dibromobutane (±) + meso 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10219-06
A10219-14
A10219-22
CAAAA10219-06EA
35.23
CAD
CAAAA10219-06
CAAAA10219-14
CAAAA10219-22
2,3-Dibromobutane (±) + meso 98+%
2,3-Dibromobutane
MDL: MFCD00000146
Beilstein Registry No.: 1718916
Purity: 98+%
CAS No.: 5408-86-6
Molecular Formula: C4H8Br2
Formula Weight: 215.93
Flash Point: 123°(253°F)
Refractive Index: 1.5126
Boiling Point: 166-167°
Beilstein Reference: 1718916
MDL No.: MFCD00000146
Formula:
H₃C(BrCH)₂CH₃ Boiling Pt: 166…167 °C Melting Pt: –24 °C Density: 1.760 Flash Pt: 123 °C(253 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000146 CAS Number: 5408-86-6 EINECS: 226-476-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...