2-Acetylbenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10216-09
A10216-18
A10216-30
CAAAA10216-09EA
89.43
CAD
CAAAA10216-09
CAAAA10216-18
CAAAA10216-30
2-Acetylbenzoic acid 98+%
2-Acetylbenzoic acid
MDL: MFCD00002475
Beilstein Registry No.: 1210557
Purity: 98+%
CAS No.: 577-56-0
Molecular Formula: C9H8O3
Formula Weight: 164.16
Odor: Odorless
Melting Point: 115-118°
Beilstein Reference: 1210557
MDL No.: MFCD00002475
Formula:
C₉H₈O₃ Boiling Pt: 110 - 112 °C (2 -rr) Melting Pt: 115…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002475 CAS Number: 577-56-0 EINECS: 209-413-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...