4-Bromo-m-xylene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dimethylbromobenzene, 1-Bromo-2,4-dimethylbenzene

A10212-14 A10212-22 A10212-36
CAAAA10212-14EA 72.05 CAD
CAAAA10212-14 CAAAA10212-22 CAAAA10212-36
4-Bromo-m-xylene 97%
4-Bromo-m-xylene

MDL: MFCD00000071


Purity: 97% CAS No.: 583-70-0 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 78°(174°F) Refractive Index: 1.5510 Boiling Point: 212-213° MDL No.: MFCD00000071

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 212…213 °C
Melting Pt: –1 °C
Density: 1.366
Flash Pt: 78 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000071
CAS Number: 583-70-0
EINECS: 209-517-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR