Tetraethylammonium tetrafluoroborate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrylammonium tetrafluoroborate

A10211-06 A10211-22 A10211-18 A10211-30
CAAAAA10211-06EA 50.09 CAD
CAAAAA10211-06 CAAAAA10211-22 CAAAA10211-18 CAAAA10211-30
Tetraethylammonium tetrafluoroborate 99%
Tetraethylammonium tetrafluoroborate

MDL: MFCD00011827 Beilstein Registry No.: 3917638


Purity: 99% CAS No.: 429-06-1 Molecular Formula: C8H20BF4N Formula Weight: 217.06 Melting Point: 370-380° dec. Beilstein Reference: 3917638 UN No.: 2811 MDL No.: MFCD00011827

Formula: C₈H₂₀BF₄N
MW: 217.06 g/mol
Melting Pt: 370…380 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011827
CAS Number: 429-06-1
EINECS: 207-055-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR