1-Adamantanol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-1-ol, Adamantan-1-ol
A10209-09
A10209-18
A10209-30
CAAAA10209-09EA
62.73
CAD
CAAAA10209-09
CAAAA10209-18
CAAAA10209-30
1-Adamantanol 99%
1-Adamantanol
MDL: MFCD00074729
Beilstein Registry No.: 1903952
Purity: 99%
CAS No.: 768-95-6
Molecular Formula: C10H16O
Formula Weight: 152.24
Melting Point: ca 245° subl.
Beilstein Reference: 1903952
Formula:
C₁₀H₁₆O Boiling Pt: 287 °C (1013 hPa) Melting Pt: ∼ 245 °C (subl.) Density: 1.06 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00074729 CAS Number: 768-95-6 EINECS: 212-202-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...