4-Chloroanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-chlorobenzoic acid

A10201-14 A10201-22 A10201-18 A10201-30
CAAAAA10201-06EA 102.84 CAD
CAAAAA10201-06 CAAAAA10201-14 CAAAAA10201-22 CAAAA10201-18 CAAAA10201-30
4-Chloroanthranilic acid 98%
4-Chloroanthranilic acid

MDL: MFCD00007778 Beilstein Registry No.: 743349


Purity: 98% CAS No.: 89-77-0 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 232° dec. Beilstein Reference: 743349

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: ∼ 232 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007778
CAS Number: 89-77-0
EINECS: 201-938-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR