L(-)-Proline 99%
Supplier: Thermo Scientific Chemicals
A10199-14
A10199-22
A10199-36
CAAAA10199-14EA
59.83
CAD
CAAAA10199-14
CAAAA10199-22
CAAAA10199-36
L(-)-Proline 99%
L(-)-Proline
MDL: MFCD00064318
Beilstein Registry No.: 80810
Fieser: 6,492 8,421 9,393 10,331 12,414
Optical Rotation: -84°C (c=1 in water)
Purity: 99%
CAS No.: 147-85-3
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Melting Point: ca 225° dec.
Beilstein Reference: 80810
Merck Reference: 14,7780
Optical Rotation: -84° (c=1 in water)
MDL No.: MFCD00064318
Formula:
C₅H₉NO₂ Boiling Pt: 245 °C (1013 hPa) Melting Pt: ∼ 225 °C (dec.) Density: 1.36 Storage Temperature: Ambient |
MDL Number:
MFCD00064318 CAS Number: 147-85-3 EINECS: 205-702-2 Merck Index: 14,7780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...