Quinaldic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
A10192-22
A10192-06
A10192-14
CAAAAA10192-22EA
1472.56
CAD
CAAAAA10192-22
CAAAA10192-06
CAAAA10192-14
Quinaldic acid 98%
Quinaldic acid
MDL: MFCD00006752
Beilstein Registry No.: 126322
Purity: 98%
CAS No.: 93-10-7
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point: 156-159°
Beilstein Reference: 126322
Merck Reference: 14,8046
MDL No.: MFCD00006752
Formula:
C₁₀H₇NO₂ Melting Pt: 156…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006752 CAS Number: 93-10-7 EINECS: 202-218-3 Merck Index: 14,8046 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...