4,4'-Bipyridyl 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10187-06
A10187-14
A10187-22
CAAAA10187-06EA
56.41
CAD
CAAAA10187-06
CAAAA10187-14
CAAAA10187-22
4,4'-Bipyridyl 98%
4,4'-Bipyridyl
MDL: MFCD00006416
Beilstein Registry No.: 113176
Purity: 98%
CAS No.: 553-26-4
Molecular Formula: C10H8N2
Formula Weight: 156.19
Melting Point: 111-114°
Boiling Point: 304-305°
Merck Reference: 14,3348
UN No.: 2811
MDL No.: MFCD00006416
Formula:
C₁₀H₈N₂ Boiling Pt: 304…305 °C Melting Pt: 111…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006416 CAS Number: 553-26-4 EINECS: 209-036-3 UN: 2811 ADR: 6.1,III Merck Index: 14,3348 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...