3-Amino-6-chloropyridazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10185-06
A10185-14
CAAAA10185-03EA
63.82
CAD
CAAAA10185-03
CAAAA10185-06
CAAAA10185-14
3-Amino-6-chloropyridazine 98%
3-Amino-6-chloropyridazine
MDL: MFCD00051506
Beilstein Registry No.: 110918
Purity: 98%
CAS No.: 5469-69-2
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Melting Point: ca 218° dec.
Beilstein Reference: 110918
Formula:
C₄H₄ClN₃ Melting Pt: ∼ 218 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051506 CAS Number: 5469-69-2 EINECS: 226-796-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...