3,5-Bis(trifluoromethyl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α,α,α,α',α',α'-Hexafluoro-3,5-xylenol

A10183-03 A10183-06 A10183-14
CAAAA10183-03EA 49.45 CAD
CAAAA10183-03 CAAAA10183-06 CAAAA10183-14
3,5-Bis(trifluoromethyl)phenol 97%
3,5-Bis(trifluoromethyl)phenol

MDL: MFCD00000386 Beilstein Registry No.: 2123670


Purity: 97% CAS No.: 349-58-6 Molecular Formula: C8H4F6O Formula Weight: 230.11 Flash Point: 110°(230°F) Refractive Index: 1.4150 Melting Point: 20-21° Boiling Point: 97-98°/50mm Beilstein Reference: 2123670 MDL No.: MFCD00000386

Formula: C₈H₄F₆O
MW: 230.11 g/mol
Boiling Pt: 97…98 °C/50 mm
Melting Pt: 20…21 °C
Density: 1.511
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000386
CAS Number: 349-58-6
EINECS: 206-488-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR