Propyl-4-hydroxybenzoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

A10180-0B A10180-0I A10180-30
CAAAA10180-0BEA 184.86 CAD
CAAAA10180-0B CAAAA10180-0I CAAAA10180-30
Propyl-4-hydroxybenzoate 99+%
Propyl 4-hydroxybenzoate

MDL: MFCD00002354 Beilstein Registry No.: 1103245


Purity: 99+% CAS No.: 94-13-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 180°(356°F) Melting Point: 95-99° Beilstein Reference: 1103245 Merck Reference: 14,7866 MDL No.: MFCD00002354

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 292
Melting Pt: 95
Flash Pt: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002354
CAS Number: 94-13-3
EINECS: 202-307-7
Merck Index: 14,7866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR