Pentyl benzoate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzoic acid pentyl ester, Amyl benzoate, Benzoic acid amyl ester

A10177-22 A10177-36
CAAAA10177-22EA 59.88 CAD
CAAAA10177-22 CAAAA10177-36
Pentyl benzoate 98+%
Pentyl benzoate

MDL: MFCD00048843


Purity: 98+% CAS No.: 2049-96-9 Molecular Formula: C12H16O2 Formula Weight: 192.26 Flash Point: 130°(266°F) Refractive Index: 1.4950 Boiling Point: 260° MDL No.: MFCD00048843

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 260 °C
Density: 0.99
Flash Pt: 130 °C(266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048843
CAS Number: 2049-96-9
EINECS: 218-077-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR