2-Tolualdehyde 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with hydroquinone (HQ) 0.1%
A10172-14
A10172-22
CAAAA10172-14EA
93.38
CAD
CAAAA10172-14
CAAAA10172-22
2-Tolualdehyde 98% stabilized
2-Tolualdehyde
MDL: MFCD00003338
Beilstein Registry No.: 605841
Purity: 98%
CAS No.: 529-20-4
Molecular Formula: C8H8O
Formula Weight: 120.15
Flash Point: 67°(152°F)
Refractive Index: 1.5470
Odor: Almond-like
Melting Point: -35°
Boiling Point: 199-200°
Beilstein Reference: 605841
Merck Reference: 14,9527
MDL No.: MFCD00003338
Formula:
C₈H₈O Boiling Pt: 199…200 °C Melting Pt: –35 °C Density: 1.037 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003338 CAS Number: 529-20-4 EINECS: 208-452-2 Merck Index: 14,9527 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...