3,4-Dimethoxybenzoic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10167-0B
A10167-18
A10167-30
CAAAAA10167-0BEA
546.68
CAD
CAAAAA10167-0B
CAAAA10167-18
CAAAA10167-30
3,4-Dimethoxybenzoic acid 99+%
3,4-Dimethoxybenzoic acid
MDL: MFCD00002500
Beilstein Registry No.: 518285
Purity: 99+%
CAS No.: 93-07-2
Molecular Formula: C9H10O4
Formula Weight: 182.18
Odor: Odorless
Melting Point: 179-183°
Beilstein Reference: 518285
Merck Reference: 14,9953
MDL No.: MFCD00002500
Formula:
C₉H₁₀O₄ Boiling Pt: 303 °C (1013 hPa) Melting Pt: 179…183 °C Density: 1.436 g/cm³ (-100 °C) Flash Pt: ≥ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002500 CAS Number: 93-07-2 EINECS: 202-215-7 Merck Index: 14,9953 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...