L(+)-Potassium sodium tartrate tetrahydrate 99%

Supplier: Thermo Scientific Chemicals

A10163-0B A10163-0I A10163-30
CAAAA10163-0BEA 278.08 CAD
CAAAA10163-0B CAAAA10163-0I CAAAA10163-30
L(+)-Potassium sodium tartrate tetrahydrate 99%
L(+)-Potassium sodium tartrate tetrahydrate

Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors


Purity: 99% CAS No.: 6381-59-5 Molecular Formula: C4H4KNaO6.4H2O Formula Weight: 282.23 (210.29anhy) Physical Form: Crystals Density: 1.77 Appearance: White Odor: Odorless pH: 6.0-8.5 Melting Point: 70-80° Solubility: Soluble in water. Insoluble in alcohol Beilstein Reference: 6113568 Merck Reference: 14,7670 Optical Rotation: +21° (c=1 in water) MDL No.: MFCD00150989


Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors

Formula: KOOC(CHOH)₂COONa·4H₂O
Melting Pt: 70…80 °C
Density: 1.77
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 14,7670
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR