3,5-Dimethyl-1H-pyrazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10157-0B
A10157-18
A10157-30
CAAAAA10157-0BEA
535.62
CAD
CAAAAA10157-0B
CAAAA10157-18
CAAAA10157-30
3,5-Dimethyl-1H-pyrazole 99%
3,5-Dimethyl-1H-pyrazole
MDL: MFCD00005243
Beilstein Registry No.: 106325
Fieser: 18,105
Purity: 99%
CAS No.: 67-51-6
Molecular Formula: C5H8N2
Formula Weight: 96.13
pH: 6.7
Melting Point: 105-108°
Boiling Point: 218°
Beilstein Reference: 106325
MDL No.: MFCD00005243
Formula:
C₅H₈N₂ Boiling Pt: 218 °C Melting Pt: 105…108 °C Density: 0.8839 g/cm³ (16 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005243 CAS Number: 67-51-6 EINECS: 200-657-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...