2-Iodoanisole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10150-36
A10150-14
A10150-22
CAAAAA10150-36EA
685.72
CAD
CAAAAA10150-36
CAAAA10150-14
CAAAA10150-22
2-Iodoanisole 99%
2-Iodoanisole
MDL: MFCD00001039
Beilstein Registry No.: 1860243
Purity: 99%
CAS No.: 529-28-2
Molecular Formula: C7H7IO
Formula Weight: 234.04
Flash Point:
Formula:
C₇H₇IO Boiling Pt: 240…242 °C Density: 1.800 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001039 CAS Number: 529-28-2 EINECS: 208-456-4 Merck Index: 14,5028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...