2,3-Dihydroxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10144-30
A10144-09
A10144-18
CAAAAA10144-30EA
2325.76
CAD
CAAAAA10144-30
CAAAA10144-09
CAAAA10144-18
2,3-Dihydroxybenzoic acid 98%
2,3-Dihydroxybenzoic acid
MDL: MFCD00002446
Beilstein Registry No.: 2209117
Purity: 98%
CAS No.: 303-38-8
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: 204-208°
Beilstein Reference: 2209117
Formula:
C₇H₆O₄ Boiling Pt: 339 °C (1013 hPa) Melting Pt: 204…208 °C Density: 1,542 Storage Temperature: Ambient |
MDL Number:
MFCD00002446 CAS Number: 303-38-8 EINECS: 206-139-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...