Ethyl-2-chlorobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A10141-14 A10141-22 A10141-36
CAAAA10141-14EA 50.09 CAD
CAAAA10141-14 CAAAA10141-22 CAAAA10141-36
Ethyl-2-chlorobenzoate 98%
Ethyl 2-chlorobenzoate

MDL: MFCD00016338 Beilstein Registry No.: 2207359


Purity: 98% CAS No.: 7335-25-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Flash Point: 110°(230°F) Refractive Index: 1.5240 Boiling Point: 243° Beilstein Reference: 2207359 MDL No.: MFCD00016338

Formula: C₉H₉ClO₂
Boiling Pt: 243 °C
Density: 1.194
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016338
CAS Number: 7335-25-3
EINECS: 230-842-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR